missing translation for 'onlineSavingsMsg'
Learn More

Fenoterol Hydrobromide, TRC

Product Code. 30393311
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393311 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393311 Supplier Toronto Research Chemicals Supplier No. TRCF24885025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenoterol Hydrobromide
CAS 1944-12-3
Purity Grade Notes HPLC
Molecular Formula C17 H21 N O4 . Br H
Quantity 25 mg
Synonym Fenoterol hydrobromide, (1RS)-1-(3,5-Dihydroxyphenyl)-2-[[(1RS)-2-(4-hydroxyphenyl)-1-methylethyl]amino]ethanol hydrobromide, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide (1:1), 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide (9CI), Benzyl alcohol, 3,5-dihydroxy-α-[[(p-hydroxy-α-methylphenethyl)amino]methyl]-, hydrobromide (8CI), 1-(3,5-Dihydroxyphenyl)-2-[2-(3-p-hydroxyphenyl)propylamino]ethanol hydrobromide, 1-(p-Hydroxyphenyl)-2-(β-3',5'-dihydroxyphenyl-β-hydroxy)ethylaminopropane hydrobromide, 3,5-Dihydroxy-α-[(p-hydroxy-α-methylphenethyl)amino]methylbenzyl alcohol hydrobromide, 3,5-Dihydroxy-α-[[(p-hydroxy-α-methylphenethyl)amino]methyl]benzyl alc. monohydrobromide, Airum, Berotec, Dosberotec, Fenoterol bromide, Fenoterol hydrobromide, Partusisten, Phenoterol hydrobromide, Th 1165a, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,R*)-(±)-, 1,3-Benzenediol, 5-[1-hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-, hydrobromide, (R*,R*)-
InChI Formula InChI=1S/C17H21NO4.BrH/c1-11(6-12-2-4-14(19)5-3-12)18-10-17(22)13-7-15(20)9-16(21)8-13;/h2-5,7-9,11,17-22H,6,10H2,1H3;1H/t11-,17+;/m1./s1
SMILES Br.C[C@H](Cc1ccc(O)cc1)NC[C@H](O)c2cc(O)cc(O)c2
IUPAC Name 5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-hydroxyphenyl)propan-2-yl]amino]ethyl]benzene-1,3-diol;hydrobromide
Molecular Weight (g/mol) 384.26
Formula Weight 383.0732
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.