missing translation for 'onlineSavingsMsg'
Learn More

Fenoterol-d6 HBr (propyl-d6-amino) (mixture of diastereomers), CDN

Product Code. 30312206
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312206 0.01 g 0.01g
30316390 0.005 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312206 Supplier CDN Supplier No. CDND79600.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenoterol-d6 Hydrobromide
CAS 1286129-04-1
Molecular Formula C17 D6 H15 N O4 . Br H
Quantity 0.01 g
Synonym 5-[2-[[1,1,1,2,3,3-Hexadeuterio-3-(4-hydroxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]benzene-1,3-diol hydrobromide, 5-[1-Hydroxy-2-[[2-(4-hydroxyphenyl)- 1-(methyl-d3)ethyl-1,2,2-d3]amino]ethyl]-1,3-benzenediol hydrobromide, 5-[2-[2-(4-Hydroxyphenyl)-1-(methyl-d3)ethyl-1,2,2-d3]amino-1-hydroxyethyl]benzene-1,3-diol hydrobromide, Fenoterol-d6 hydrobromide, rac-Fenoterol-D6 hydrobromide (1-(methyl-D3)ethyl-1,2,2-D3)
InChI Formula InChI=1 S/C17H21NO4.BrH/c1-11(6-12-2-4-14(19)5-3-12)18-10-17(22)13-7-15(20)9-16(21)8-13;/h2-5,7-9,11,17-22 H,6,10H2,1H3;1 H/i1D3,6D2,11 D;
SMILES Br.[2 H]C([2 H])([2 H])C([2 H])(NCC(O)c1cc(O)cc(O)c1)C([2 H])([2 H])c2ccc(O)cc2
IUPAC Name 5-[2-[[1,1,1,2,3,3-hexadeuterio-3-(4-hydroxyphenyl)propan-2-yl]amino]-1-hydroxyethyl]benzene-1,3-diol;hydrobromide
Molecular Weight (g/mol) 390.302
Formula Weight 389.111 g/mol
Percent Purity 97 atom % D, min 97% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.