missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenoprofen, TRC
Specifications
Specifications
| Chemical Name or Material | Fenoprofen |
| CAS | 29679-58-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H14 O3 |
| Quantity | 50 mg |
| Synonym | Benzeneacetic acid, α-methyl-3-phenoxy-, Hydratropic acid, m-phenoxy- (8CI), α-Methyl-3-phenoxybenzeneacetic acid, (±)-2-(3-Phenoxyphenyl)propionic acid, (±)-Fenoprofen, (±)-m-Phenoxyhydratropic acid, 2-(3-Phenoxyphenyl)-1-propanoic acid, 2-(3-Phenoxyphenyl)propanoic acid, 2-(3-Phenoxyphenyl)propionic acid, 2-(m-Phenoxyphenyl)propionic acid, 3-Phenoxyhydratropic acid, Fenoprofen, Lilly 53858, dl-2-(3-Phenoxyphenyl)propionic acid, m-Phenoxyhydratropic acid |
| InChI Formula | InChI=1S/C15H14O3/c1-11(15(16)17)12-6-5-9-14(10-12)18-13-7-3-2-4-8-13/h2-11H,1H3,(H,16,17) |
| SMILES | CC(C(=O)O)c1cccc(Oc2ccccc2)c1 |
| IUPAC Name | 2-(3-phenoxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 242.27 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?