missing translation for 'onlineSavingsMsg'
Learn More

Fenoldopam mesylate, MedChemExpress

Product Code. 16418598
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418598 10 mg 10mg
16448436 100 mg 100mg
16424467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16418598 Supplier Medchem Express Supplier No. HYB0735A
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fenoldopam(SKF 82526) mesylate is a drug and synthetic benzazepine derivative which acts as a selective D1 receptor partial agonist.

Fenoldopam(SKF 82526) mesylate is a drug and synthetic benzazepine derivative which acts as a selective D1 receptor partial agonist.

Specifications

Chemical Name or Material Fenoldopam mesylate
CAS 67227-57-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H20ClNO6S
Quantity 10 mg
Synonym Fenoldopam methanesulfonate SKF-82526 mesylate
Solubility Information DMSO : ≥ 36 mg/mL (89.58 mM) ∣H2O : 10 mg/mL (24.88 mM; Need ultrasonic)
SMILES OC1=C(O)C=C2C(C3=CC=C(O)C=C3)CNCCC2=C1Cl.CS(=O)(O)=O
Molecular Weight (g/mol) 401.86
Formula Weight 401.86
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.