missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid, TRC

Product Code. 17931395
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931395 10 mg 10mg
17973105 100 mg 100mg
17912815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931395 Supplier Toronto Research Chemicals Supplier No. TRCF24865010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42017-89-0
Molecular Formula C17 H15 Cl O4
Molecular Weight (g/mol) 318.75
Synonym Fenofibric Acid, Propanoic acid, 2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-, 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid, 2-[4'-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, 2-[p-(p-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, FNF acid, LF 153, LF 178 acid, NSC 281318, Procetofenic acid, α-1081
IUPAC Name 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)
Formula Weight 318.0659
Percent Purity >95%
Chemical Name or Material Fenofibric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.