missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric acid, MedChemExpress

Product Code. 16472269
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472269 1 g 1g
16421058 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16472269 Supplier Medchem Express Supplier No. HYB0760
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fenofibric acid, an active metabolite of fenofibrate, is a PPAR activitor, with EC50s of 22.4 μM, 1.47 μM, and 1.06 μM for PPARα, PPARγ and PPARδ, respectively; Fenofibric acid also inhibits COX-2 enzyme activity, with an IC50 of 48 nM.

Fenofibric acid, an active metabolite of fenofibrate, is a PPAR activitor, with EC50s of 22.4 μM, 1.47 μM, and 1.06 μM for PPARα, PPARγ and PPARδ, respectively

Specifications

Chemical Name or Material Fenofibric acid
CAS 42017-89-0
Color White
Purity Grade Notes Research
Molecular Formula C17H15ClO4
Quantity 1 g
Synonym FNF acid
Solubility Information DMSO : ≥ 100 mg/mL (313.73 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(C)(OC1=CC=C(C(C2=CC=C(Cl)C=C2)=O)C=C1)C(O)=O
Molecular Weight (g/mol) 318.75
Formula Weight 318.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.