missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid Ethyl Ester, TRC

Product Code. 30008114
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008114 1 g 1g
30006866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008114 Supplier Toronto Research Chemicals Supplier No. TRCF2486701G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42019-08-9
Molecular Formula C19 H19 Cl O4
Molecular Weight (g/mol) 346.8
Synonym Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate, Fenofibrate Imp. E (EP)
IUPAC Name ethyl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H19ClO4/c1-4-23-18(22)19(2,3)24-16-11-7-14(8-12-16)17(21)13-5-9-15(20)10-6-13/h5-12H,4H2,1-3H3
Formula Weight 346.0972
Chemical Name or Material Ethyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.