missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid Acyl-Beta-D-glucuronide Allyl Ester, TRC

Product Code. 30401387
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401387 50 mg 50mg
30403190 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401387 Supplier Toronto Research Chemicals Supplier No. TRCF24866550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenofibric Acid Acyl-beta-D-glucuronide Allyl Ester
Analyte or Component Names Fenofibric Acid Acyl-beta-D-glucuronide Allyl Ester
Shipping Condition Room Temperature
Molecular Formula C26H27ClO10
Synonym 1-[2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate] β-D-Glucopyranuronic Acid Allyl Ester
InChI Formula InChI=1S/C26H27ClO10/c1-4-13-34-23(32)22-20(30)19(29)21(31)24(35-22)36-25(33)26(2,3)37-17-11-7-15(8-12-17)18(28)14-5-9-16(27)10-6-14/h4-12,19-22,24,29-31H,1,13H2,2-3H3/t19-,20-,21+,22-,24-/m0/s1
Solution Type Neat
SMILES O=C(O[C@@H]1O[C@H](C(OCC=C)=O)[C@@H](O)[C@H](O)[C@H]1O)C(C)(C)OC2=CC=C(C(C3=CC=C(Cl)C=C3)=O)C=C2
IUPAC Name allyl (2S,3S,4S,5R,6S)-6-((2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 534.94
Quantity 50 mg
Formula Weight 534.13
Show More Show Less
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.