missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid Acyl-Beta-D-glucuronide Allyl Ester, TRC

Product Code. 30403190
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403190 5 mg 5mg
30401387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30403190 Supplier Toronto Research Chemicals Supplier No. TRCF2486655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenofibric Acid Acyl-beta-D-glucuronide Allyl Ester
Analyte or Component Names Fenofibric Acid Acyl-beta-D-glucuronide Allyl Ester
Shipping Condition Room Temperature
Molecular Formula C26H27ClO10
Synonym 1-[2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoate] β-D-Glucopyranuronic Acid Allyl Ester
InChI Formula InChI=1S/C26H27ClO10/c1-4-13-34-23(32)22-20(30)19(29)21(31)24(35-22)36-25(33)26(2,3)37-17-11-7-15(8-12-17)18(28)14-5-9-16(27)10-6-14/h4-12,19-22,24,29-31H,1,13H2,2-3H3/t19-,20-,21+,22-,24-/m0/s1
Solution Type Neat
SMILES O=C(O[C@@H]1O[C@H](C(OCC=C)=O)[C@@H](O)[C@H](O)[C@H]1O)C(C)(C)OC2=CC=C(C(C3=CC=C(Cl)C=C3)=O)C=C2
IUPAC Name allyl (2S,3S,4S,5R,6S)-6-((2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoyl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 534.94
Quantity 5 mg
Formula Weight 534.13
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.