missing translation for 'onlineSavingsMsg'
Learn More

Fenofibric Acid Acyl-Beta-D-glucuronide (~90%), TRC

Product Code. 30007085
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007085 5 mg 5mg
30010264 10 mg 10mg
30008927 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007085 Supplier Toronto Research Chemicals Supplier No. TRCF2486605MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60318-63-0
Molecular Formula C23 H23 Cl O10
Molecular Weight (g/mol) 494.88
Synonym Fenofibric Acid Acyl-Beta-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H23ClO10/c1-23(2,22(31)33-21-18(28)16(26)17(27)19(32-21)20(29)30)34-14-9-5-12(6-10-14)15(25)11-3-7-13(24)8-4-11/h3-10,16-19,21,26-28H,1-2H3,(H,29,30)/t16-,17-,18+,19-,21-/m0/s1
Formula Weight 494.098
Chemical Name or Material Fenofibric Acid Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.