missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fenofibric acid

Product Code. 17231202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17231202 5 g 5g
17221202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231202 Supplier Thermo Scientific Chemicals Supplier No. 466170050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42017-89-0
Molecular Formula C17H15ClO4

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: .. Other solubilities: practically insoluble in chloroform, soluble in acetone and methanol
Melting Point 187.0°C
Formula Weight 318.75
InChI Key MQOBSOSZFYZQOK-UHFFFAOYSA-N
SMILES CC(C)(OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1)C(O)=O
IUPAC Name 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
Molecular Weight (g/mol) 318.75
Chemical Name or Material Fenofibric acid
compliance-icons compliance-icons
Product Identifier
  • Fenofibric acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.