missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate, MedChemExpress

Product Code. 16477298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477298 10 g 10g
16459807 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477298 Supplier Medchem Express Supplier No. HY17356
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fenofibrate is a selective PPARα agonist with an EC50 of 30 μM. Fenofibrate also inhibits human cytochrome P450 isoforms, with IC50s of 0.2, 0.7, 9.7, 4.8 and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively.

Fenofibrate is a selective PPARα agonist with an EC50 of 30 μM. Fenofibrate also inhibits human cytochrome P450 isoforms, with IC50s of 0.2, 0.7, 9.7, 4.8 and 142.1 μM for CYP2C19, CYP2B6, CYP2C9, CYP2C8, and CYP3A4, respectively.

Specifications

Chemical Name or Material Fenofibrate
CAS 49562-28-9
Color White
Purity Grade Notes Research
Molecular Formula C20H21ClO4
Quantity 10 g
Solubility Information DMSO : 250 mg/mL (692.85 mM; Need ultrasonic)
SMILES CC(C)(OC1=CC=C(C(C2=CC=C(Cl)C=C2)=O)C=C1)C(OC(C)C)=O
Molecular Weight (g/mol) 360.83
Formula Weight 360.83
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.