missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Fenobam
GREENER_CHOICE

Product Code. 30001442
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001442

Brand: Tocris Bioscience™ 2386/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective mGlu5 negative allosteric modulator (NAM)

Potent and selective non-competitive mGlu5 negative allosteric modulator (NAM) that also displays inverse agonist properties; blocks mGlu5 constitutive activity in vitro (IC50 = 87 nM). Acts at an allosteric modulatory site shared with MPEP and binds the mGlu5 receptor with Kd values of 54 and 31 nM for rat and human receptors respectively. Displays anxiolytic activity following oral administration in vivo; also exhibits analgesic properties.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57653-26-6
Molecular Formula C11H11ClN4O2

Specifications

Quantity 10 mg
Formula Weight 266.69
Purity >99%
Molecular Weight (g/mol) 266.69
Chemical Name or Material Fenobam
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.