missing translation for 'onlineSavingsMsg'
Learn More

Fenclozic Acid, TRC

Product Code. 30394873
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394873 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394873 Supplier Toronto Research Chemicals Supplier No. TRCF2475301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenclozic Acid
CAS 17969-20-9
Molecular Formula C11H8ClNO2S
Quantity 1 g
Synonym 2-(p-Chlorophenyl)-4-thiazoleacetic Acid, 2-(4-Chlorophenyl)thiazol-4-ylacetic Acid, 2-(p-Chlorophenyl)thiazol-4-ylacetic Acid, Acidum Fenclozicum, ICI 54450, Mialex, Myalex, 2-(2-(4-Chlorophenyl)thiazol-4-yl)acetic Acid
InChI Formula InChI=1S/C11H8ClNO2S/c12-8-3-1-7(2-4-8)11-13-9(6-16-11)5-10(14)15/h1-4,6H,5H2,(H,14,15)
SMILES O[C@@H]1C[C@@H]2[C@H](CC1)[C@H]3[C@@H](CC2)[C@@H]4[C@@](CC3)(C)[C@@H](O)CC4
IUPAC Name 2-[2-(4-chlorophenyl)-1,3-thiazol-4-yl]acetic acid
Molecular Weight (g/mol) 253.7
Formula Weight 252.9964
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.