missing translation for 'onlineSavingsMsg'
Learn More

Fenclonine, MedChemExpress

Product Code. 16412339
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412339 Supplier Medchem Express Supplier No. HYB1368
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fenclonine is a potent and irreversible inhibitor of tryptophan (Trp) hydroxylase, the rate-limiting enzyme in serotonin biosynthesis.

Fenclonine is a potent and irreversible inhibitor of tryptophan (Trp) hydroxylase, the rate-limiting enzyme in serotonin biosynthesis.

Specifications

Chemical Name or Material Fenclonine
CAS 7424-00-2
Color White
Purity Grade Notes Research
Molecular Formula C9H10ClNO2
Quantity 1 g
Synonym 4-Chloro-DL-phenylalanine PCPA CP-10188
Solubility Information DMSO : 100 mg/mL (500.93 mM; ultrasonic and adjust pH to 2 with HCl) ∣H2O : 25 mg/mL (125.23 mM; ultrasonic and adjust pH to 10 with NaOH)
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O
Molecular Weight (g/mol) 199.63
Formula Weight 199.63
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.