missing translation for 'onlineSavingsMsg'
Learn More

Fenclofenac, TRC

Product Code. 30402753
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402753 250 mg
30392072 50 mg
30406155 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402753 Supplier Toronto Research Chemicals Supplier No. TRCF247505250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34645-84-6
Molecular Formula C14 H10 Cl2 O3
Molecular Weight (g/mol) 297.13
Synonym Benzeneacetic acid, 2-(2,4-dichlorophenoxy)-, Fenclofenac, Flenac, R 67408, RX 67408, [o-(2,4-Dichlorophenoxy)phenyl]acetic acid
IUPAC Name 2-[2-(2,4-dichlorophenoxy)phenyl]acetic acid
SMILES OC(=O)Cc1ccccc1Oc2ccc(Cl)cc2Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H10Cl2O3/c15-10-5-6-13(11(16)8-10)19-12-4-2-1-3-9(12)7-14(17)18/h1-6,8H,7H2,(H,17,18)
Formula Weight 296.0007
Percent Purity >95%
Chemical Name or Material Fenclofenac
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.