missing translation for 'onlineSavingsMsg'
Learn More

Fenclofenac Acyl-Beta-D-Glucuronide Allyl Ester, TRC

Product Code. 30394740
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394740 250 mg 250mg
30407730 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394740 Supplier Toronto Research Chemicals Supplier No. TRCF247525250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenclofenac Acyl-beta-D-Glucuronide Allyl Ester
Analyte or Component Names Fenclofenac Acyl-beta-D-Glucuronide Allyl Ester
Shipping Condition Room Temperature
Molecular Formula C23H22Cl2O9
Synonym 1-[2-(2,4-Dichlorophenoxy)benzeneacetate]β-D-Glucopyranuronic Acid Propenyl Ester
InChI Formula InChI=1S/C23H22Cl2O9/c1-2-9-31-22(30)21-19(28)18(27)20(29)23(34-21)33-17(26)10-12-5-3-4-6-15(12)32-16-8-7-13(24)11-14(16)25/h2-8,11,18-21,23,27-29H,1,9-10H2/t18-,19-,20+,21-,23+/m0/s1
Solution Type Neat
SMILES O=C(O[C@@H]1O[C@H](C(OCC=C)=O)[C@@H](O)[C@H](O)[C@H]1O)CC2=CC=CC=C2OC3=CC=C(Cl)C=C3Cl
IUPAC Name allyl (2S,3S,4S,5R,6S)-6-(2-(2-(2,4-dichlorophenoxy)phenyl)acetoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 513.32
Quantity 250 mg
Formula Weight 512.06
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.