missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone, 98+%

Product Code. 11318176
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318176 50 g 50g
11328176 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318176 Supplier Thermo Scientific Alfa Aesar Supplier No. L15151.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7787-20-4 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Melting Point 3°C to 5°C
Density 0.945
Boiling Point 192°C to 194°C
Flash Point 73°C (163°F)
Refractive Index 1.463
Quantity 50 g
Beilstein 2042710
Optical Rotation −52° (Neat)
Formula Weight 152.24
Percent Purity ≥98%
Chemical Name or Material (-)-Fenchone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.