missing translation for 'onlineSavingsMsg'
Learn More

Fenbutrazate, TRC

Product Code. 30135374
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135374 100 mg 100mg
30007756 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135374 Supplier Toronto Research Chemicals Supplier No. TRCF247250100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4378-36-3
Molecular Formula C23 H29 N O3
Molecular Weight (g/mol) 367.48
Synonym Butyric acid, 2-phenyl-, 2-(3-methyl-2-phenylmorpholino)ethyl ester (6CI,7CI,8CI), 2-(3-Methyl-2-phenylmorpholino)ethyl 2-phenylbutyrate, 2-(3-Methyl-2-phenylmorpholino)ethyl α-phenylbutyrate, 2-Phenylbutyric acid 2-(3-methyl-2-phenylmorpholino)ethyl ester, Fenbutrazate, Fenbutrazatum, Nethanol, Phenbutrazate, Phenbutrazatum
IUPAC Name 2-(3-methyl-2-phenylmorpholin-4-yl)ethyl 2-phenylbutanoate
SMILES CCC(C(=O)OCCN1CCOC(C1C)c2ccccc2)c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H29NO3/c1-3-21(19-10-6-4-7-11-19)23(25)27-17-15-24-14-16-26-22(18(24)2)20-12-8-5-9-13-20/h4-13,18,21-22H,3,14-17H2,1-2H3
Formula Weight 367.2147
Percent Purity >95%
Chemical Name or Material Fenbutrazate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.