missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenbendazole Sulphone, Mikromol™
Descripción
Fenbendazole Sulphone
Especificaciones
Especificaciones
| Chemical Name or Material | Fenbendazole Sulfone |
| CAS | 54029-20-8 |
| Analyte or Component Names | Fenbendazole Sulfone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl [5-(Phenylsulphonyl)-1H-benzimidazol-2-yl]carbamate, Fenbendazole Sulphone, Oxfendazole Imp. B (EP), Carbamic acid, [5-(phenylsulfonyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Fenbendazole sulfone, HOE 5151, Methyl [5-(phenylsulfonyl)-1H-benzimidazol-2-yl]carbamate, Oxfendazole sulfone, Oxfendazole Impurity B |
| InChI Formula | InChI=1S/C15H13N3O4S/c1-22-15(19)18-14-16-12-8-7-11(9-13(12)17-14)23(20,21)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19) |
| SMILES | COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)(=O)c3ccccc3 |
| IUPAC Name | methyl N-[5-(benzenesulfonyl)-1H-benzimidazol-2-yl]carbamate |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?