missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole Sulfoxide, TRC

Product Code. 30402554
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402554 50 mg -
30391466 10 mg 10mg
30395346 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402554 Supplier Toronto Research Chemicals Supplier No. TRCF24679050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15 H13 N3 O3 S
Molecular Weight (g/mol) 315.35
Synonym Fenbendazole Sulphoxide, Oxfendazole, Fenbendazole Sulfoxide
IUPAC Name methyl N-[5-(benzenesulfinyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H13N3O3S/c1-21-15(19)18-14-16-12-8-7-11(9-13(12)17-14)22(20)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)
Formula Weight 315.0678
Chemical Name or Material Oxfendazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.