missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole Sulfone, TRC

Product Code. 17913875
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913875 10 mg 10mg
17913415 5 mg 5mg
17932375 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913875 Supplier Toronto Research Chemicals Supplier No. TRCF24677010MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54029-20-8
Molecular Formula C15 H13 N3 O4 S
Molecular Weight (g/mol) 331.35
Synonym Carbamic acid, [5-(phenylsulfonyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), Fenbendazole sulfone, HOE 5151, Methyl [5-(phenylsulfonyl)-1H-benzimidazol-2-yl]carbamate, Oxfendazole sulfone
IUPAC Name methyl N-[5-(benzenesulfonyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)(=O)c3ccccc3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H13N3O4S/c1-22-15(19)18-14-16-12-8-7-11(9-13(12)17-14)23(20,21)10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)
Formula Weight 331.0627
Percent Purity >95%
Chemical Name or Material Fenbendazole Sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.