missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole, Mikromol™

Product Code. 18042780
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18042780 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18042780 Supplier Mikromol Supplier No. MM0518.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fenbendazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenbendazole
CAS 43210-67-9
Analyte or Component Names Fenbendazole
Shipping Condition Room Temperature
Molecular Formula C15 H13 N3 O2 S
Synonym Fenbendazole, Methyl [5-(phenylsulfanyl)-1H-benzimidazol-2-yl]carbamate, 2-(Methoxycarbonylamino)-5-(phenylthio)benzimidazole, Axilur, Febendazole, Fenbendazol, Fenbendazole, Fenbion, HOE 881, Methyl 5-(phenylthio)-2-benzimidazolecarbamate, Methyl [5-(phenylthio)-1H-benzimidazol-2-yl]carbamate, Panacur, Safe-Guard, Vigisol, Febantel Impurity C, Oxfendazole Impurity A
InChI Formula InChI=1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)
SMILES COC(=O)Nc1nc2cc(Sc3ccccc3)ccc2[nH]1
IUPAC Name methyl N-(5-phenylsulfanyl-1H-benzimidazol-2-yl)carbamate
Molecular Weight (g/mol) 299.35
Quantity 250 mg
Formula Weight 299.0728 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.