missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole, MedChemExpress

Product Code. 16442126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442126 500 mg 500mg
16408296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442126 Supplier Medchem Express Supplier No. HYB0413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fenbendazole is a broad spectrum benzimidazole anthelmintic used against gastrointestinal parasites.

Fenbendazole is a broad spectrum benzimidazole anthelmintic used against gastrointestinal parasites.

Specifications

Chemical Name or Material Fenbendazole
Physical Form Solid
CAS 43210-67-9
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C15H13N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 10 mg/mL (33.41 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(OC)NC1=NC2=CC=C(SC3=CC=CC=C3)C=C2N1
Molecular Weight (g/mol) 299.35
Formula Weight 299.35
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.