missing translation for 'onlineSavingsMsg'
Learn More

Fenbendazole D3 (methyl D3) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30319492
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30319492

Brand: Dr. Ehrenstorfer DREA13446010AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenbendazole-d3
CAS 1228182-47-5
Solvent or Matrix Acetonitrile
Molecular Formula C15H10D3N3O2S
Quantity 1 mL
Synonym Trideuteriomethyl N-(5-phenylsulfanyl-1H-benzimidazol-2-yl)carbamate, Trideuteriomethyl N-(6-phenylsulfanyl-1H-benzimidazol-2-yl)carbamate
InChI Formula InChI=1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)/i1D3
SMILES [2H]C([2H])([2H])OC(=O)Nc1nc2ccc(Sc3ccccc3)cc2[nH]1
IUPAC Name trideuteriomethyl N-(6-phenylsulfanyl-1H-benzimidazol-2-yl)carbamate
Molecular Weight (g/mol) 302.37
Formula Weight 302.0917 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.