missing translation for 'onlineSavingsMsg'
Learn More

Fenazaquin, TRC

Product Code. 30397797
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397797 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397797 Supplier Toronto Research Chemicals Supplier No. TRCF24574050MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenazaquin
CAS 120928-09-8
Purity Grade Notes HPLC
Molecular Formula C20 H22 N2 O
Quantity 50 mg
Synonym 4-[2-[4-(1,1-Dimethylethyl)phenyl]ethoxy]quinazoline, EL 436, Fenaza, Fenazachin, Fenazaquin, GWN 1708, Magister, Magus, Phenazaquin, Pride Ultra, XDE 436
InChI Formula InChI=1S/C20H22N2O/c1-20(2,3)16-10-8-15(9-11-16)12-13-23-19-17-6-4-5-7-18(17)21-14-22-19/h4-11,14H,12-13H2,1-3H3
SMILES CC(C)(C)c1ccc(CCOc2ncnc3ccccc23)cc1
IUPAC Name 4-[2-(4-tert-butylphenyl)ethoxy]quinazoline
Molecular Weight (g/mol) 306.4
Formula Weight 306.1732
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.