missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenamiphos 90%, TRC
Specifications
Specifications
| Chemical Name or Material | Fenamiphos |
| CAS | 22224-92-6 |
| Molecular Formula | C13 H22 N O3 P S |
| Quantity | 500 mg |
| Synonym | Phosphoramidic acid, (1-methylethyl)-, ethyl 3-methyl-4-(methylthio)phenyl ester (9CI), Phosphoramidic acid, isopropyl-, ethyl 4-(methylthio)-m-tolyl ester (8CI), B 68138, BAY 68138, Bayer 68138, Ethyl 3-methyl-4-(methylthio)phenyl (1-methylethyl)phosphoramidate, Ethyl 4-(methylthio)-m-tolyl isopropylphosphoramidate, Fenamifos, Fenamiphos, Fenamithion, Nemacur, Nemacur P, Phenamiphos, Phosphoramidic acid, N-(1-methylethyl)-, ethyl 3-methyl-4-(methylthio)phenyl ester |
| InChI Formula | InChI=1S/C13H22NO3PS/c1-6-16-18(15,14-10(2)3)17-12-7-8-13(19-5)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15) |
| SMILES | CCOP(=O)(NC(C)C)Oc1ccc(SC)c(C)c1 |
| IUPAC Name | N-[ethoxy-(3-methyl-4-methylsulfanylphenoxy)phosphoryl]propan-2-amine |
| Molecular Weight (g/mol) | 303.36 |
| Formula Weight | 303.1058 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?