missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14768-1-AP-150UL
This item is not returnable.
View return policy
Description
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5′ flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.Specifications
| FEN1 | |
| Polyclonal | |
| Unconjugated | |
| FEN1 | |
| DNase IV, FEN 1, FEN1, Flap endonuclease 1, flap structure endonuclease 1, hFEN 1, Maturation factor 1, MF1, RAD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14156, 2237, 84490 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P39748, P39749, Q5XIP6 | |
| Fen1 | |
| FEN1 Fusion Protein Ag6552 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction