missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FEN1 Monoclonal Antibody (A1B12)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA542534

Product Code. 17719153

  • £429.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: MCF-7 cell lysate, K562 cell lysate, Daudi cell lysate, PC-12 cell lysate, NIH/3T3 cell lysate, human tonsil tissue, human liver carcinoma tissue, human breast carcinoma tissue, K562. Subcellular Location: Nucleolus, nucleoplasm. Mitochondrion.

FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FEN1
Monoclonal
2 mg/mL
TBS with 50% glycerol and 0.05% sodium azide; pH 7.4
P39748, P39749, Q5XIP6
FEN1
Recombinant protein within human FEN1 aa 150-380.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
A1B12
Unconjugated
FEN1
AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2
Mouse
Protein G
RUO
14156, 2237, 84490
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.