missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Antibody (SAIC-21C-4) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
£190.00 - £494.00
Specifications
| Antigen | FEN-1 |
|---|---|
| Clone | SAIC-21C-4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631555
|
Novus Biologicals
NBP3-20095-0.025mg |
0.025 mg |
£190.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670995
|
Novus Biologicals
NBP3-20095-0.2mg |
0.2 mg |
£494.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FEN-1 Monoclonal antibody specifically detects FEN-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| FEN-1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| The KLH-cojugated peptide 'SIEEIVR', derived from the Flap endonuclease 1 involved in DNA replication and repair. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| SAIC-21C-4 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| PBS | |
| 2237 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title