missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEN-1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-320AF647
This item is not returnable.
View return policy
Description
FEN-1 Polyclonal antibody specifically detects FEN-1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| FEN-1 | |
| Polyclonal | |
| Western Blot | |
| EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2 | |
| The immunogen recognized by this antibody maps to a region between residues 75 and 125 of human Flap Structure Endonuclease 1 using the numbering given in entry NP_004102.1 (GeneID 2237). | |
| 0.1 mL | |
| Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases | |
| 2237 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction