Learn More
Value Plastics Female Luer Thread Union Fittings
Shop All Cole-Parmer Products
Click to view available options
Material (Seal):
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer Thread |
| Material (Seal) | Polypropylene |