Learn More
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Product Code. 30053983 Shop All Cole-Parmer Products
Change view
Click to view available options
Thread Size:
1/4-28 UNF(M)
1/4-28 UNF(M) Bottom Sealing
10-32 UNF(M)
Inlet:
Female Luer with 1/4 in. Hex
Female Luer with 5/16 in. Hex
Material (Seal):
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Inlet Material (Seal) Unit Size
30053983 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Polypropylene Pack of 25
30053201 1/4-28 UNF(M) Bottom Sealing Female Luer with 5/16 in. Hex Polypropylene Pack of 25
30053683 10-32 UNF(M) Female Luer with 1/4 in. Hex Nylon Pack of 25
30054148 1/4-28 UNF(M) Bottom Sealing Female Luer with 5/16 in. Hex Nylon Pack of 25
30053992 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Nylon Pack of 25
30054264 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Polycarbonate Pack of 25
30054098 10-32 UNF(M) Female Luer with 1/4 in. Hex Polycarbonate Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30053983 Supplier Value Plastics Supplier No. SFTLL6005
  • ONLINE EXCLUSIVE PRICE
    £39.20
    £36.24 / Pack of 25
    Save £2.96
    7% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Luer fittings allow for a simple yet secure connection
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Packaging Quantity 25/Pk.
Inlet Female Luer with 5/16 in. Hex
Thread Size 1/4-28 UNF(M)
Material (Seal) Polypropylene