Learn More
Value Plastics Female Luer, Panel Mount 1/4-28 UNF to 200 Series Barb
Product Code. p-200105070 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Product Code. 30053104
Brand: Value Plastics 5012040
Only null left
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 1/8 in.
Material Nylon
Packaging Quantity 25/Pk.
Inlet Female Luer with Panel Mount, Hose Barb
Always test the application to verify the performance of the interconnection.