Learn More
Cole-Parmer™ Female Luer Lock NPT
Click to view available options
Thread Size:
1/4 in. NPT
Unit Size:
Each
Description
- Easy to disassemble for cleaning
- Ideal for harsh chemical environments
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel (316) |
| Port Size | 1/4 in. NPT female luer |
| Thread Size | 1/4 in. NPT |