Learn More
Cytiva Female Luer Lock Connector for ÄKTA Systems
Cytiva Female Luer Lock Connector can be used with Autosampler A-900 and A-905, an integral component of numerous ÄKTA chromatography systems.
Brand: Cytiva 18-1120-38
Code : GX
Additional Details : Weight : 0.00200kg
Description
Cytiva Female Luer Lock Connector can be used with numerous ÄKTA liquid chromatography systems, including: ÄKTAexplorer™, ÄKTApurifier™, ÄKTAfplc™, or ÄKTAmicro™ systems.Specifications
Female Luer Lock Connector | |
1 Ea. | |
Autosampler A-900 and A-905, Used in Numerous ÄKTA Systems, Including: ÄKTAexplorer, ÄKTApurifier, Ä |
Luer lok, female | |
Clear |