missing translation for 'onlineSavingsMsg'
Learn More

Colder Products Company Female Luer Genderless MicroCNX Connector

Product Code. 30968691
Change view
Click to view available options
Material:
Polycarbonate
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30968691 Polycarbonate Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30968691 Supplier Colder Products Company Supplier No. CNX170LFHT
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MicroCNX™ Connectors are the only aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small-format biomanufacturing assemblies.

  • The MicroCNX line of connectors is engineered specifically for the challenging conditions of biological media transfer in bioprocessing, cell therapy, and gene therapy applications.
  • The connector comes in a range of tubing sizes; a luer connection option to create a sterile, closed connection on an existing luer connection; and the ULT version is ideal for assemblies for cryogenic freezing cassettes or connection to PVC tubing.
  • Finish: Molded White
  • Pressure Range: Up to 43.5 psi, 3 bar
  • Free Floating
  • Temperature Range: 34°F to 104°F (1°C to 40°C)
  • Seal Options: Silicone, platinum-cured, USP Class VI, ADCF
  • Non-Valved

Specifications

Autoclavable Autoclavable
Material Polycarbonate
Sterility Non-sterile
Color White, Blue
Product Type Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.