Learn More
Value Plastics Female Luer to 200 Series Barb
Product Code. 30053083 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/18 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Material unitSize
30053083 1/8 in. Nylon Pack of 25
30053501 1/16 in. Nylon Pack of 25
30054140 1/16 in. Polycarbonate Pack of 25
30054270 1/16 in. Polypropylene Pack of 25
30054126 5/32 in. Nylon Pack of 25
30053387 5/32 in. Polypropylene Pack of 25
30053941 3/32 in. Polycarbonate Pack of 25
30052979 1/8 in. Polycarbonate Pack of 25
30053616 3/16 in. Polypropylene Pack of 25
30054218 3/32 in. Polypropylene Pack of 25
30054301 3/16 in. Nylon Pack of 25
30054266 5/32 in. Polycarbonate Pack of 25
30053368 3/32 in. Nylon Pack of 25
30053483 1/18 in. Polypropylene Pack of 25
30054005 3/16 in. Polycarbonate Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 30053083 Supplier Value Plastics Supplier No. 5012038
Only null left
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 1/8 in.
Material Nylon
Packaging Quantity 25/Pk.
Inlet Female Luer x Hose Barb
Always test the application to verify the performance of the interconnection.