Learn More
Value Plastics Female Luer to 200 Series Barb
Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/18 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 1/18 in. |
| Material | Polypropylene |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer x Hose Barb |
Always test the application to verify the performance of the interconnection.