Learn More
FEM1C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531869
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (86%), Xenopus laevis (91%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.Specifications
FEM1C | |
Polyclonal | |
Unconjugated | |
FEM1C | |
2610312A07Rik; 3632443A22Rik; AI646567; EUROIMAGE686608; EUROIMAGE783647; fem-1 homolog C; fem-1 homolog c (C. elegans); fem-1 homolog c (C.elegans); FEM1A; FEM1c; FEM1-gamma; feminization 1 homolog a; feminization 1 homolog c; KIAA1785; Protein fem-1 homolog C; protein fem-1 homolog C pseudogene; protein fem-1 homolog C-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96JP0 | |
FEM1C | |
Recombinant fragment corresponding to a region within amino acids 1 and 228 of Human FEM1C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |