missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FEM1C Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563794
This item is not returnable.
View return policy
Description
Immunogen sequence: NLLIKSGAHF DATNLHKQTA SDLLDEKEIA KNLIQPINHT TLQCLAARVI VNHRIYYKGH I Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.
Specifications
| FEM1C | |
| Polyclonal | |
| Unconjugated | |
| FEM1C | |
| 2610312A07Rik; 3632443A22Rik; AI646567; EUROIMAGE686608; EUROIMAGE783647; fem-1 homolog C; fem-1 homolog c (C. elegans); fem-1 homolog c (C.elegans); FEM1A; FEM1c; FEM1-gamma; feminization 1 homolog a; feminization 1 homolog c; KIAA1785; Protein fem-1 homolog C; protein fem-1 homolog C pseudogene; protein fem-1 homolog C-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JP0 | |
| FEM1C | |
| Recombinant protein corresponding to Human FEM1C. Recombinant protein control fragment (Product #RP-100149). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction