missing translation for 'onlineSavingsMsg'
Learn More

FEM1B Antibody (4B12), Novus Biologicals™

Product Code. 18352289 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352289 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352289 Supplier Novus Biologicals Supplier No. H00010116M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FEM1B Monoclonal antibody specifically detects FEM1B in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEM1B
Applications ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 4B12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_056137
Gene Alias DKFZp451E0710, F1AA, F1A-ALPHA, FEM-1 (C.elegans) homolog b, fem-1 homolog b (C. elegans), FEM1b, FEM1-beta, FEM-1-like death receptor binding protein, Fem-1-like death receptor-binding protein alpha, Fem-1-like in apoptotic pathway protein alpha, FIAA, KIAA0396, protein fem-1 homolog B
Host Species Mouse
Immunogen FEM1B (NP_056137.1, 401 a.a. ~ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TVKAPDIECVLRCSVLEIEQSMNRVKNISDADVHNAMDNYECNLYTFLYLVCISTKTQCSEEDQCKINKQIYNLIHLDPRTREGFTLLHL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 10116
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.