missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEM1A Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-36956
This item is not returnable.
View return policy
Description
FEM1A Polyclonal specifically detects FEM1A in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| FEM1A | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp762M136, EPRAP, fem-1 homolog a (C. elegans), FEM1a, FEM1-alpha, Prostaglandin E receptor 4-associated protein, protein fem-1 homolog A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55527 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_061178.1 | |
| FEM1A | |
| Peptide with sequence C-GFIPEDLEAFIELH corresponding to C-Terminus according to NP_061178.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the C Terminus. Cross reacts with Human. Expected from sequence similarity: Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction