missing translation for 'onlineSavingsMsg'
Learn More

Fellutamide B (Mixture of Diastereomers, >80%), TRC

Product Code. 30388983
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388983 2.5 mg 2.5mg
30394817 5 mg -
30401859 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388983 Supplier Toronto Research Chemicals Supplier No. TRCF1380002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138752-26-8
Molecular Formula C27H49N5O7
Molecular Weight (g/mol) 555.71
Synonym N2-[(3R)-3-Hydroxy-1-oxododecyl]-L-asparaginyl-N1-(1-formyl-3-methylbutyl)-L-glutamamide, (2S)-2-((S)-4-Amino-2-((R)-3-hydroxydodecanamido)-4-oxobutanamido)-N1-(4-methyl-1-oxopentan-2-yl)pentanediamide
IUPAC Name (2S)-2-[[(2S)-4-amino-2-[[(3R)-3-hydroxydodecanoyl]amino]-4-oxobutanoyl]amino]-N-(4-methyl-1-oxopentan-2-yl)pentanediamide
SMILES CCCCCCCCC[C@@H](O)CC(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H](CCC(=O)N)C(=O)NC(CC(C)C)C=O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C27H49N5O7/c1-4-5-6-7-8-9-10-11-20(34)15-25(37)31-22(16-24(29)36)27(39)32-21(12-13-23(28)35)26(38)30-19(17-33)14-18(2)3/h17-22,34H,4-16H2,1-3H3,(H2,28,35)(H2,29,36)(H,30,38)(H,31,37)(H,32,39)/t19?,20-,21+,22+/m1/s1
Formula Weight 555.3632
Chemical Name or Material Fellutamide B (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.