missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Feline Leukemia Virus Monoclonal Antibody (YCH)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MIF2801
This item is not returnable.
View return policy
Description
MIF2801 targets Feline Leukemia Virus in ELISA, IP, and RIA applications and shows reactivity with Virus samples. The MIF2801 immunogen is viral coat antigen. MIF2801 was formerly sold as a Seradyn product.
Feline leukemia virus (FeLV) is a retrovirus that infects cats. As a retrovirus, FeLV is transmitted as an RNA virus but the RNA is reverse transcribed into DNA. Retroviruses contradict the Central dogma of molecular biology. FeLV is usually transmitted between infected cats when the transfer of saliva or nasal secretions is involved. If not defeated by the animal's immune system, the virus can be lethal. The disease caused by this virus is a form of cancer of blood cells called lymphocytes (a leukemia).
Specifications
| Feline Leukemia Virus | |
| Monoclonal | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| YCH | |
| Unconjugated | |
| FLV | |
| Viral coat antigen. | |
| 1 mg | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction