missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline IFN-gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB764
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Feline samples. It is validated for Immunocytochemistry.
Specifications
| IFN-gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3458 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 770626 | |
| Immunocytochemistry 8-25 ug/mL | |
| P46402 | |
| IFNG | |
| E. coli-derived recombinant feline IFN-gamma Gln24-Lys167 Accession # P46402 | |
| 100 μg | |
| Primary | |
| Detects feline IFN-gamma in direct ELISAs. | |
| Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction