missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline CXCR4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4287
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Feline samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| CXCR4 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| P56498 | |
| CXCR4 | |
| HEK293 human embryonic kidney cell line transfected with feline CXCR4 Met1-Ser353 Accession # P56498 | |
| 100 μg | |
| Primary | |
| Detects feline CXCR4. Stains feline CXCR4 transfectants but not irrelevant transfectants. | |
| Feline | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 374606 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7852 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction