missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Feline CD4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2597-SP
This item is not returnable.
View return policy
Description
CD4 Polyclonal specifically detects CD4 in Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| CD4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD4 | |
| Mouse myeloma cell line NS0-derived recombinant feline CD4 | |
| 25 μg | |
| Primary | |
| Detects feline CD4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant canine CD4 is observed and 5% cross-reactivity with recombinant human CD4 and recombinant mouse CD4 is observed. | |
| Feline | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 920 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction