missing translation for 'onlineSavingsMsg'
Learn More

Felbinac, TRC

Product Code. 30408542
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408542 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408542 Supplier Toronto Research Chemicals Supplier No. TRCF23110010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Felbinac
CAS 5728-52-9
Molecular Formula C14 H12 O2
Quantity 10 g
Synonym Felbinac, (Biphenyl-4-yl)acetic acid
InChI Formula InChI=1S/C14H12O2/c15-14(16)10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H,10H2,(H,15,16)
SMILES OC(=O)Cc1ccc(cc1)c2ccccc2
IUPAC Name 2-(4-phenylphenyl)acetic acid
Molecular Weight (g/mol) 212.24
Formula Weight 212.0837
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.