missing translation for 'onlineSavingsMsg'
Learn More

Felbinac Isopropyl Ester, Mikromol™

Product Code. 18081050
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18081050 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081050 Supplier Mikromol Supplier No. MM3501.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Felbinac Isopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Felbinac Isopropyl Ester
CAS 1286423-88-8
Analyte or Component Names Felbinac Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C17 H18 O2
Certifications/Compliance ISO 9001
Synonym Felbinac Isopropyl Ester, [1,1'-Biphenyl]-4-acetic acid 1-methylethyl ester
InChI Formula InChI=1S/C17H18O2/c1-13(2)19-17(18)12-14-8-10-16(11-9-14)15-6-4-3-5-7-15/h3-11,13H,12H2,1-2H3
SMILES CC(C)OC(=O)Cc1ccc(cc1)c2ccccc2
IUPAC Name propan-2-yl 2-(4-phenylphenyl)acetate
Molecular Weight (g/mol) 254.32
Quantity 100 mg
Formula Weight 254.1307 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.